Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SSX2 Antibody (OTI4D10), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP274368G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274368G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274368G Supplier Novus Biologicals Supplier No. NBP274368G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SSX2 Monoclonal specifically detects SSX2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen SSX2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI4D10
Conjugate DyLight 488
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Cancer/testis antigen 5.2, cancer/testis antigen family 5, member 2a, CT5.2a, HD21, HOM-MEL-40, MGC119055, MGC15364, MGC3884, protein SSX2, sarcoma, synovial, X-chromosome-related 2, SSX, SSX2A, SSX2B, synovial sarcoma, X breakpoint 2B, synovial sarcoma, X breakpoint 2CT5.2, Tumor antigen HOM-MEL-40
Gene Symbols SSX2
Host Species Mouse
Immunogen Full-length protein expressed in 293T cell transfected with human SSX2 expression vector.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 6757.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.