Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ST2/IL-33R Antibody (003), PerCP, Novus Biologicals™
SDP

Catalog No. NB308169 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB308169 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB308169 Supplier Novus Biologicals Supplier No. NBP289270PCP
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

ST2/IL-33R Monoclonal antibody specifically detects ST2/IL-33R in Human samples. It is validated for ELISA

Specifications

Antigen ST2/IL-33R
Applications ELISA
Classification Monoclonal
Clone 003
Conjugate PerCP
Dilution ELISA
Formulation PBS
Gene Alias DER4ST2growth stimulation-expressed, FIT-1, IL33R, interleukin 1 receptor-like 1, interleukin 1 receptor-related protein, interleukin-1 receptor-like 1, MGC32623, Protein ST2, ST2L, ST2V, T1homolog of mouse growth stimulation-expressed
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human ST2/IL-33R (Met 1-Phe 328; Accession#: NP_003847.2).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis
Primary or Secondary Primary
Gene ID (Entrez) 9173
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.