Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ST2/IL-33R Antibody, Novus Biologicals™
SDP

Catalog No. NBP224706 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224706 0.1 mg
NBP224706SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224706 Supplier Novus Biologicals Supplier No. NBP224706
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ST2/IL-33R Polyclonal specifically detects ST2/IL-33R in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen ST2/IL-33R
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 3-5 ug/ml
Formulation 0.2 mls PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. Q01638
Gene Alias DER4ST2growth stimulation-expressed, FIT-1, IL33R, interleukin 1 receptor-like 1, interleukin 1 receptor-related protein, interleukin-1 receptor-like 1, MGC32623, Protein ST2, ST2L, ST2V, T1homolog of mouse growth stimulation-expressed
Gene Symbols IL1RL1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 207-238 of human ST2V was used as immunogen, GenBank no BAA85894.
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Angiogenesis
Primary or Secondary Primary
Gene ID (Entrez) 9173
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoform1 and isoform 2), monkey and rat, and 91% homologous in chicken.
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.