Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ST2/IL-33R Antibody (ST33868), Biotin, Novus Biologicals™
SDP

Catalog No. NB172332 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB172332 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB172332 Supplier Novus Biologicals Supplier No. NBP280040B
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

ST2/IL-33R Monoclonal antibody specifically detects ST2/IL-33R in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry

Specifications

Antigen ST2/IL-33R
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone ST33868
Conjugate Biotin
Formulation PBS
Gene Alias DER4ST2growth stimulation-expressed, FIT-1, IL33R, interleukin 1 receptor-like 1, interleukin 1 receptor-related protein, interleukin-1 receptor-like 1, MGC32623, Protein ST2, ST2L, ST2V, T1homolog of mouse growth stimulation-expressed
Host Species Mouse
Immunogen Recombinant human soluble ST2/IL-33R.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis
Primary or Secondary Primary
Gene ID (Entrez) 9173
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.