Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ST2/IL-33R, Mouse anti-Human, Clone: ST33868, Novus Biologicals™
SDP

Catalog No. NB013158 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB013158 50 μg
NB013157 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB013158 Supplier Novus Biologicals Supplier No. NBP28004050UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ST2/IL-33R Monoclonal specifically detects ST2/IL-33R in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen ST2/IL-33R
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone ST33868
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation 0.2 um-filtered solution in PBS, pH 7.4
Gene Alias DER4ST2growth stimulation-expressed, FIT-1, IL33R, interleukin 1 receptor-like 1, interleukin 1 receptor-related protein, interleukin-1 receptor-like 1, MGC32623, Protein ST2, ST2L, ST2V, T1homolog of mouse growth stimulation-expressed
Gene Symbols IL1RL1
Host Species Mouse
Immunogen Recombinant human ST2/IL-33R (soluble).
Purification Method Protein G purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9173
Test Specificity Recognizes human ST2/IL-33R.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.