Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ST2 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 7A2A7, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | ST2 |
---|---|
Clone | 7A2A7 |
Concentration | 1 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6245
![]() ![]() |
Proteintech Group Inc
601121IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
ST2 is a member of the interleukin 1 receptor family. Studies of ST2 in mouse suggested that this receptor can be induced by proinflammatory stimuli, and may be involved in the function of helper T cells. ST2 belongs to the interleukin-1 receptor family with three Ig-like C2-type (immunoglobulin-like) domains and a TIR domain. ST2 is a receptor for interleukin-33 (IL-33) which upon stimulation recruits the signaling molecules MYD88, IRAK1, IRAK4, and TRAF6, followed by phosphorylation of MAPK3/ERK1 and/or MAPK1/ERK2, MAPK14, and MAPK8. Reports also suggest its possible role in helper T-cell function as well as in cell growth. Three isoforms of ST2 have been reported where isoform C is a membrane form and isoform B is a secreted form. ST2 may be induced by proinflammatory stimuli, and may also be found in serum of autoimmune disease. Different isoforms are restricted to different tissues while high expression of this protein is evident in kidney, lung, placenta, stomach, skeletal muscle, colon and small intestine.Specifications
ST2 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
DER4, FIT 1, IL1RL1, interleukin 1 receptor like 1, Protein ST2, ST2, ST2L, ST2V, T1 | |
Il1rl1 | |
IgG1 | |
Caprylic Acid Precipitation | |
Antibody |
7A2A7 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Pig, Rat | |
P14719, Q01638, Q62611 | |
100127134, 17082, 25556, 9173 | |
ST2 Fusion Protein Ag2555 | |
Primary | |
-20°C | |
IL1RL1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title