Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ST3 beta-Gal alpha-2,3-Sialyltransferase 1/ST3GAL1/SIAT4A Antibody, Novus Biologicals™
SDP

Catalog No. NBP131449 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP131449 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP131449 Supplier Novus Biologicals Supplier No. NBP131449
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ST3 beta-Gal alpha-2,3-Sialyltransferase 1/ST3GAL1/SIAT4A Polyclonal specifically detects ST3 beta-Gal alpha-2,3-Sialyltransferase 1/ST3GAL1/SIAT4A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen ST3 beta-Gal alpha-2,3-Sialyltransferase 1/ST3GAL1/SIAT4A
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.98mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation PBS (pH 7), 1% BSA, 20% Glycerol with 0.025% Proclin 300
Gene Alias Alpha 2,3-ST 1, CMP-N-acetylneuraminate-beta-galactosamide-alpha-2,3-sialyltransferase 1, DKFZp666E036, DKFZp779K2051, EC 2.4.99, EC 2.4.99.4, Gal-beta-1,3-GalNAc-alpha-2,3-sialyltransferase, Gal-NAc6S, Sialyltransferase 4A, sialyltransferase 4A (beta-galactosidase alpha-2,3-sialytransferase), sialyltransferase 4A (beta-galactoside alpha-2,3-sialyltransferase), sialyltransferase 4A (beta-galactoside alpha-2,3-sialytransferase), SIAT4, SIAT4A, SIAT4-A, SIATFLFLJ36548, ST3 beta-galactoside alpha-2,3-sialyltransferase 1, ST3Gal I, ST3GalA.1MGC9183, ST3GalI, ST3GalIA, ST3GalIA1,Beta-galactoside alpha-2,3-sialyltransferase 1, ST3OST3GalA
Gene Symbols ST3GAL1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human SIAT4A. The exact sequence is proprietary.
Molecular Weight of Antigen 39 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6482
Test Specificity Tested in human. Not yet tested in any other species.
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.