Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ST3 beta-Gal alpha-2,3-Sialyltransferase 2/ST3GAL2 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB320598 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB320598 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB320598 Supplier Novus Biologicals Supplier No. NBP298478F

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Polyclonal Antibody

ST3 beta-Gal alpha-2,3-Sialyltransferase 2/ST3GAL2 Polyclonal antibody specifically detects ST3 beta-Gal alpha-2,3-Sialyltransferase 2/ST3GAL2 in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen ST3 beta-Gal alpha-2,3-Sialyltransferase 2/ST3GAL2
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate FITC
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias Alpha 2,3-ST 2, Beta-galactoside alpha-2,3-sialyltransferase 2, beta-galactoside alpha-2,3-sialytransferase, CMP-N-acetylneuraminate-beta-galactosamide-alpha-2,3-sialyltransferase 2, EC 2.4.99.4, EC 2.4.99.7, Gal-beta-1,3-GalNAc-alpha-2,3-sialyltransferase, Gal-NAc6S, Sialyltransferase 4B, sialyltransferase 4B (beta-galactosidase alpha-2,3-sialytransferase), SIAT4B, SIAT4-B, ST3 beta-galactoside alpha-2,3-sialyltransferase 2, ST3GalA.2ST3Gal II, ST3GALII
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human ST3 beta-Gal alpha-2,3-Sialyltransferase 2/ST3GAL2 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6483
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.