Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ST3GAL5 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | ST3GAL5 |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
ST3GAL5 Polyclonal specifically detects ST3GAL5 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
ST3GAL5 | |
Polyclonal | |
Rabbit | |
Signal Transduction | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
8869 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:NKTTIRMTYPEGAPLSDLEYYSNDLFVAVLFKSVDFNWLQAMVKKETLPFWVRLFFWKQVAEKIPLQPKHFRILNPVIIKETAFDILQYSEPQSRFWG | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Human | |
alpha 2,3-sialyltransferase V, CMP-NeuAc:lactosylceramide alpha-2,3-sialyltransferase, EC 2.4.99.9, Ganglioside GM3 synthase, GM3 synthase, lactosylceramide alpha-2,3-sialyltransferase, Sialyltransferase 9, sialyltransferase 9 (CMP-NeuAc:lactosylceramide alpha-2,3-sialyltransferase;GM3 synthase), SIATGM3S, ST3 beta-galactoside alpha-2,3-sialyltransferase 5, ST3Gal V, ST3GALV, ST3GalVSIAT9SATI | |
ST3GAL5 | |
IgG | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title