Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ST6 Gal Sialyltransferase 1/ST6GAL1/CD75 Antibody (001), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB317285 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB317285 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB317285 Supplier Novus Biologicals Supplier No. NBP289922APC
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

ST6 Gal Sialyltransferase 1/ST6GAL1/CD75 Monoclonal antibody specifically detects ST6 Gal Sialyltransferase 1/ST6GAL1/CD75 in Human samples. It is validated for ELISA

Specifications

Antigen ST6 Gal Sialyltransferase 1/ST6GAL1/CD75
Applications ELISA
Classification Monoclonal
Clone 001
Conjugate APC
Dilution Sandwich ELISA Capture
Formulation PBS
Gene Alias B-cell antigen CD75, beta-galactoside alpha-2,6-sialyltransferase 1, CMP-N-acetylneuraminate beta-galactosamide alpha-2,6-sialyltransferase, CMP-N-acetylneuraminate-beta-galactosamide-alpha-2,6-sialyltransferase 1, EC 2.4.99.1, MGC48859, Sialyltransferase 1, sialyltransferase 1 (beta-galactoside alpha-2,6-sialyltransferase), sialyltransferase 1 (beta-galactoside alpha-2,6-sialytransferase), SIAT1Alpha 2,6-ST 1, ST6 beta-galactosamide alpha-2,6-sialyltranferase 1, ST6Gal I, ST6GalI, ST6N
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human ST6 Gal Sialyltransferase 1/ST6GAL1/CD75 (Uniprot#: P15907; Lys27-Cys406).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Golgi Apparatus Markers
Primary or Secondary Primary
Gene ID (Entrez) 6480
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.