Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ST6 Gal Sialyltransferase 1/ST6GAL1/CD75 Rabbit anti-Human, Clone: 36, Novus Biologicals™
SDP

Catalog No. NB021312 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB021312 100 μg
NB021313 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB021312 Supplier Novus Biologicals Supplier No. NBP289923100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

ST6 Gal Sialyltransferase 1/ST6GAL1/CD75 Monoclonal specifically detects ST6 Gal Sialyltransferase 1/ST6GAL1/CD75 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen ST6 Gal Sialyltransferase 1/ST6GAL1/CD75
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 36
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 1:100-1:500
Formulation 0.2 um filtered solution in PBS
Gene Alias B-cell antigen CD75, beta-galactoside alpha-2,6-sialyltransferase 1, CMP-N-acetylneuraminate beta-galactosamide alpha-2,6-sialyltransferase, CMP-N-acetylneuraminate-beta-galactosamide-alpha-2,6-sialyltransferase 1, EC 2.4.99.1, MGC48859, Sialyltransferase 1, sialyltransferase 1 (beta-galactoside alpha-2,6-sialyltransferase), sialyltransferase 1 (beta-galactoside alpha-2,6-sialytransferase), SIAT1Alpha 2,6-ST 1, ST6 beta-galactosamide alpha-2,6-sialyltranferase 1, ST6Gal I, ST6GalI, ST6N
Host Species Rabbit
Immunogen Recombinant Human ST6 Gal Sialyltransferase 1/ST6GAL1/CD75 protein
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Golgi Apparatus Markers
Primary or Secondary Primary
Gene ID (Entrez) 6480
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.