Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ST6 Gal Sialyltransferase 2/ST6GAL2 Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB050097 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB050097 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB050097 Supplier Novus Biologicals Supplier No. NBP297963H

Rabbit Polyclonal Antibody

ST6 Gal Sialyltransferase 2/ST6GAL2 Polyclonal specifically detects ST6 Gal Sialyltransferase 2/ST6GAL2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen ST6 Gal Sialyltransferase 2/ST6GAL2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate HRP
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias Alpha 2,6-ST 2, beta-galactoside alpha-2,6-sialyltransferase 2, beta-galactoside alpha-2,6-sialyltransferase II, CMP-N-acetylneuraminate-beta-galactosamide-alpha-2,6-sialyltransferase 2, EC 2.4.99, EC 2.4.99.1, FLJ30711, FLJ37730, FLJ38334, hST6Gal II, KIAA1877St6gal2, Sialyltransferase 2, sialyltransferase 2 (monosialoganglioside sialyltransferase), SIAT2, ST6 beta-galactosamide alpha-2,6-sialyltranferase 2, ST6Gal II, ST6GALII
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human ST6 Gal Sialyltransferase 2/ST6GAL2 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84620
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.