Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ST6 GalNAc alpha-2,6-sialyltransferaseV/ST6GALNAC5 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB320590 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB320590 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB320590 Supplier Novus Biologicals Supplier No. NBP298479APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ST6 GalNAc alpha-2,6-sialyltransferaseV/ST6GALNAC5 Polyclonal antibody specifically detects ST6 GalNAc alpha-2,6-sialyltransferaseV/ST6GALNAC5 in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen ST6 GalNAc alpha-2,6-sialyltransferaseV/ST6GALNAC5
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate APC
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias alpha-N-acetylneuraminyl 2,3-betagalactosyl-1,3)-N-acetyl galactosaminidealpha-2,6-sialyltransferase E, EC 2.4.99, EC 2.4.99.-, EC 2.4.99.7, GalNAc alpha-2,6-sialyltransferase V, GD1 alpha synthase, MGC3184, sialyltransferase 7 (alpha-N-acetylneuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase) E, sialyltransferase 7((alpha-N-acetylneuraminyl-2,3-beta-galactosyl-1,3)-N-acetyl galactosaminidealpha-2,6-sialyltransferase) E, Sialyltransferase 7E, SIAT7-E, SIAT7Ealpha-N-acetylgalactosaminide alpha-2,6-sialyltransferase V, ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminidealpha-2,6-sialyltransferase 5, ST6 neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminidealpha-2,6-sialyltransferase 5, ST6GalNAc V, ST6GalNAcValpha-N-acetylgalactosaminide alpha-2,6-sialyltransferase 5
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human ST6 GalNAc alpha-2,6-sialyltransferaseV/ST6GALNAC5 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Epitope Tags
Primary or Secondary Primary
Gene ID (Entrez) 81849
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.