Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ST6 Sialyltransferase 2/ST6GALNAC2 Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB082075 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB082075 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB082075 Supplier Novus Biologicals Supplier No. NBP298353AF350
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ST6 Sialyltransferase 2/ST6GALNAC2 Polyclonal specifically detects ST6 Sialyltransferase 2/ST6GALNAC2 in Mouse samples. It is validated for ELISA.

Specifications

Antigen ST6 Sialyltransferase 2/ST6GALNAC2
Applications ELISA
Classification Polyclonal
Conjugate Alexa Fluor 350
Formulation 50 mM Sodium Borate
Gene Alias (alpha-N-acetylneuraminyl-2,3-beta-galactosyl-1,3)-N-acetyl galactosaminide B, alpha-N-acetylgalactosaminide alpha-2,6-sialyltransferase 2, EC 2.4.99.-, FLJ45660, GalNAc alpha-2,6-sialyltransferase II, SAITL1, sialyltransferase 7((alpha-N-acetylneuraminyl-2,3-beta-galactosyl-1,3)-N-acetyl galactosaminide B, sialyltransferase 7((alpha-N-acetylneuraminyl-2,3-beta-galactosyl-1,3)-N-acetyl galactosaminidealpha-2,6-sialyltransferase), sialyltransferase 7((alpha-N-acetylneuraminyl-2,3-beta-galactosyl-1,3)-N-acetyl galactosaminidealpha-2,6-sialyltransferase) B, Sialyltransferase 7B, sialyltransferase-like 1, SIAT7, SIAT7B, SIAT7-B, SIATL1, ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminidealpha-2,6-sialyltransferase 2, ST6GalNAc II, ST6GalNAcII, ST6GalNAII, SThM, STHM(alpha-N-acetylneuraminyl-2,3-beta-galactosyl-1,3)-N-acetyl galactosaminidealpha-2,6-sialyltransferase
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant ST6 Sialyltransferase 2/ST6GALNAC2 (P70277; Ser28-Arg373).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10610
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.