Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ST6 Sialyltransferase 6/ST6GALNAC6 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen ST6 Sialyltransferase 6/ST6GALNAC6
Dilution Western Blot 1.0 ug/ml
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB123359
SDP
View Documents
Novus Biologicals
NBP309229100UL
100 μg
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

ST6 Sialyltransferase 6/ST6GALNAC6 Polyclonal specifically detects ST6 Sialyltransferase 6/ST6GALNAC6 in Human samples. It is validated for Western Blot.
Specifications

Specifications

ST6 Sialyltransferase 6/ST6GALNAC6
Western Blot
Unconjugated
Rabbit
Human
alpha-N-acetylgalactosaminide alpha-2,6-sialyltransferase 6, CMP-NeuAC:(beta)-N-acetylgalactosaminide (alpha)2,6-sialyltransferase member VI, EC 2.4.99, EC 2.4.99.-, EC 2.4.99.7, GalNAc alpha-2,6-sialyltransferase VI, hST6GalNAc VI, RP11-203J24.3, Sialyltransferase 7F, sialytransferase 7 ((alpha-N-acetylneuraminyl 2,3-betagalactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialytransferase) F, SIAT7F, SIAT7-F, ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminidealpha-2,6-sialyltransferase 6, ST6GalNAc VI, ST6GalNAcVI
The immunogen is a synthetic peptide directed towards the N terminal region of human ST6 Sialyltransferase 6/ST6GALNAC6 (NP_038471). Peptide sequence ALITILILYSSNSANEVFHYGSLRGRSRRPVNLKKWSITDGYVPILGNKT
Affinity purified
Western Blot 1.0 ug/ml
Polyclonal
Purified
RUO
PBS buffer, 2% sucrose
30815
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.