Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ST8 alpha-2,8-Sialyltransferase 4/ST8SIA4 Antibody, Novus Biologicals™
SDP

Catalog No. NBP213393 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP213393 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP213393 Supplier Novus Biologicals Supplier No. NBP213393
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ST8 alpha-2,8-Sialyltransferase 4/ST8SIA4 Polyclonal specifically detects ST8 alpha-2,8-Sialyltransferase 4/ST8SIA4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen ST8 alpha-2,8-Sialyltransferase 4/ST8SIA4
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias Alpha-2,8-sialyltransferase 8D, CMP-N-acetylneuraminate-poly-alpha-2,8-sialyl transferase, CMP-N-acetylneuraminate-poly-alpha-2,8-sialyltransferase, EC 2.4.99, EC 2.4.99.-, Polysialyltransferase-1, PST1MGC61459, PSTMGC34450, sialyltransferase 8 (alpha-2, 8-polysialytransferase) D, Sialyltransferase 8D, Sialytransferase St8Sia IV, SIAT8D, SIAT8-D, ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 4, ST8SiaIV, ST8SIA-IV
Gene Symbols ST8SIA4
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to the amino acids: RTEEHQETQLIGDGELSLSRSLVNSSDKIIRKAGSSIFQHNVEGWKINSSLVLEIRKNILRFLDAERDVSVVKSSFKPGDVIHYVLD
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 7903
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.