Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ST8 alpha-2,8-Sialyltransferase 8A/ST8SIA1/Ganglioside GD3 Antibody, Novus Biologicals™
SDP

Catalog No. NB10075632 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10075632 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10075632 Supplier Novus Biologicals Supplier No. NB10075632
Add to Cart
Add to Cart

Chicken Polyclonal Antibody

ST8 alpha-2,8-Sialyltransferase 8A/ST8SIA1/Ganglioside GD3 Polyclonal specifically detects ST8 alpha-2,8-Sialyltransferase 8A/ST8SIA1/Ganglioside GD3 in Human samples. It is validated for Western Blot.

Specifications

Antigen ST8 alpha-2,8-Sialyltransferase 8A/ST8SIA1/Ganglioside GD3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS with No Preservative
Gene Accession No. Q6PZN5
Gene Alias Alpha-2,8-sialyltransferase 8A, alpha-N-acetylneuraminide alpha-2,8-sialyltransferase, disialoganglioside (GD3) synthase, Ganglioside GD3 synthase, Ganglioside GT3 synthase, ganglioside-specific alpha-2,8-polysialyltransferase, GD3 synthase), GD3S, sialyltransferase 8 (alpha-N-acetylneuraminate: alpha-2,8-sialytransferase, GD3synthase) A, Sialyltransferase 8A, sialyltransferase 8A (alpha-N-acetylneuraminate: alpha-2,8-sialyltransferase, Sialytransferase St8Sia I, SIAT8A, SIAT8-A, SIAT8EC 2.4.99.8, ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 1, ST8SiaI
Gene Symbols ST8SIA1
Host Species Chicken
Immunogen The immunogen for this antibody was a sequence from 4a.a. to 18a.a. from ST8SIA1(a 356a.a. long protein)
Purification Method Affinity Purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Neuronal Stem Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 6489
Test Specificity Ganglioside GD3
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.