Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ST8SIA1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | ST8SIA1 |
---|---|
Concentration | 0.27 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-4616
![]() ![]() |
Proteintech Group Inc
249181AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Gangliosides are membrane-bound glycosphingolipids containing sialic acid. Ganglioside GD3 is known to be important for cell adhesion and growth of cultured maligt cells. The protein encoded by this gene is a type II membrane protein that catalyzes theSpecifications
ST8SIA1 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q64687, Q92185 | |
20449, 6489 | |
ST8SIA1 Fusion Protein Ag20429 | |
Primary | |
-20°C | |
ST8SIA1 |
0.27 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Alpha 2,8 sialyltransferase 8A, Ganglioside GD3 synthase, Ganglioside GT3 synthase, GD3S, Sialyltransferase 8A, Sialytransferase St8Sia I, SIAT8, SIAT8 A, SIAT8A, ST8SIA1, ST8SiaI | |
ST8SIA1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title