Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ST8SIA5 Antibody, Novus Biologicals™
SDP

Catalog No. NBP230530 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
25ul
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP230530 0.1 mL
NB437548 25ul
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP230530 Supplier Novus Biologicals Supplier No. NBP230530
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ST8SIA5 Polyclonal specifically detects ST8SIA5 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin).

Specifications

Antigen ST8SIA5
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:500 - 1:1000
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. O15466
Gene Alias alpha-2,8-sialyltransferase 8E, EC 2.4.99.-, MGC119670, Sialyltransferase 8E, sialyltransferase 8E (alpha-2, 8-polysialytransferase), Sialytransferase St8Sia V, SIAT8-E, SIAT8EMGC119671, ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 5, ST8SiaV
Gene Symbols ST8SIA5
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: SGLYITHHYYDNVKPRPGFHAMPSEIFNFLHLHSRGILRVHTGTCS
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29906
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.