Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STAG2 Antibody, Novus Biologicals™
SDP

Catalog No. NB10060946 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10060946 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10060946 Supplier Novus Biologicals Supplier No. NB10060946
Add to Cart
Add to Cart

Goat Polyclonal Antibody

STAG2 Polyclonal specifically detects STAG2 in Human samples. It is validated for Immunohistochemistry, Peptide ELISA, Western Blot.

Specifications

Antigen STAG2
Applications Immunohistochemistry, Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.01 ug/ml, Immunohistochemistry 1:10 - 1:500, Peptide ELISA Detection limit 1:128000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_001036214.1
Gene Alias bA517O1.1, cohesin subunit SA-2, DKFZp686P168, SA-2, SA2FLJ25871, SCC3 homolog 2, stromal antigen 2DKFZp781H1753
Gene Symbols STAG2
Host Species Goat
Immunogen Peptide with sequence C-REPKRLRPEDS corresponding to internal region according to NP_001036214.1, NP_001036215.1 NP_001036216.1 NP_006594.3.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 10735
Test Specificity This antibody is expected to recognize isoform a (NP_001036214.1 and NP_001036215.1) and isoform b (NP_001036216.1 and NP_006594.3).
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.