Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STAG3 Antibody, Novus Biologicals™
SDP

Catalog No. NBP106084 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP106084 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP106084 Supplier Novus Biologicals Supplier No. NBP106084
Add to Cart
Add to Cart

Goat Polyclonal Antibody

STAG3 Polyclonal specifically detects STAG3 in Human, Mouse, Rat samples. It is validated for Peptide ELISA.

Specifications

Antigen STAG3
Applications ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Peptide ELISA Detection limit 1:32000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_058660.2
Gene Alias cohesin subunit SA-3, SCC3 homolog 3, stromal antigen 3stromalin 3, stromalin-3
Gene Symbols STAG3
Host Species Goat
Immunogen Peptide with sequence C-KHYNKFYEDYGD corresponding to internal region according to Uniprot Mouse NP_058660.2.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 10734
Test Specificity The immunizing peptide was designed based on the Mouse protein sequence with one residue of a diference from the human sequence.
Target Species Human, Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.