Description
- For 1/16 in. tubing on all 10-32 ports
- 1/4 in. head on all nuts
- Ferrules not included, must be purchased separately
Specifications
Specifications
| Max. Pressure | 20,000 psi |
| Port Style | 10-32 |
| For Use With (Application) | 1/16 in. Tubing |
| Material | Stainless Steel |
| Type | Valco-Style Nuts (Medium) |
| Quantity | 5/Pk. |