Learn More
Restek™ Stainless Steel Fittings
SDP
Catalog No. 06611711 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Type:
Rheodyne-Style Ferrules (Cone-Style)
Rheodyne-Style Ferrules (Stepped-Style)
Rheodyne-Style Nuts (Long)
Rheodyne-Style Nuts (Medium)
Rheodyne-Style Nuts (Short)
Valco Ferrule
Valco-Style Nuts (Long)
Valco-Style Nuts (Medium)
Valco-Style Nuts (Short)
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
06-611-711 Valco Ferrule
06-611-713 Rheodyne-Style Nuts (Medium)
06-611-714 Rheodyne-Style Nuts (Long)
06-611-716 Rheodyne-Style Ferrules (Cone-Style)
06-611-715 Rheodyne-Style Ferrules (Stepped-Style)
06-611-708 Valco-Style Nuts (Short)
06-611-709 Valco-Style Nuts (Medium)
06-611-710 Valco-Style Nuts (Long)
06-611-712 Rheodyne-Style Nuts (Short)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 06-611-711 Supplier Restek™ Supplier No. 27730
Add to Cart
Add to Cart
Ideal for connecting stainless steel tubing in UHPLC systems
  • For 1/16 in. tubing on all 10-32 ports
  • 1/4 in. head on all nuts
  • Ferrules not included, must be purchased separately
Specifications
Port Style 10-32
For Use With (Application) 1/16 in. Tubing
Material Stainless Steel
Type Valco Ferrule
Quantity 5/Pk.