Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STAM 1, 2 Antibody - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB018485 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
0.5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018485 0.1 mg
NBP130592 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018485 Supplier Novus Biologicals Supplier No. NBP1305920.1MG
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

STAM 1, 2 Polyclonal specifically detects STAM 1, 2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen STAM 1, 2
Applications Western Blot, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Formulation Sterile water with No Preservative
Gene Alias DKFZp686J2352, HSE1 homolog, signal transducing adapter molecule 1, signal transducing adaptor molecule (SH3 domain and ITAM motif) 1, STAM1STAM-1
Gene Symbols STAM
Host Species Sheep
Immunogen A synthetic peptide from the internal part of human STAM 1, 2 (STAM-1, STAM-2) conjugated to an immunogenic carrier protein was used as the immunogen. The antigen is homologous in many species including rat, mouse, zebra fish and xenopus.
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8027
Test Specificity Appears to be specific for STAM1 & 2.
Reconstitution Reconstitute with 0.5 ml sterilized water.
Target Species Mouse, Rat, Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.