Learn More
Description
Specifications
Specifications
| Antigen | STAM 1, 2 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Western Blot 10-50 ug/ml, Immunohistochemistry 10-50 ug/ml, Immunohistochemistry-Paraffin 10-50 ug/ml, Immunohistochemistry-Frozen 10-50 ug/ml |
| Formulation | Lyophilized from PBS (1x) with No Preservative |
| Gene Alias | DKFZp686J2352, HSE1 homolog, signal transducing adapter molecule 1, signal transducing adaptor molecule (SH3 domain and ITAM motif) 1, STAM1STAM-1 |
| Gene Symbols | STAM |
| Host Species | Sheep |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
