Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STAM-1 Antibody (29C678), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB10056533V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056533V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10056533V Supplier Novus Biologicals Supplier No. NB10056533V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

STAM-1 Monoclonal specifically detects STAM-1 in Human samples. It is validated for Western Blot.

Specifications

Antigen STAM-1
Applications Western Blot
Classification Monoclonal
Clone 29C678
Conjugate DyLight 405
Dilution Western Blot
Gene Accession No. Q92783
Gene Alias DKFZp686J2352, HSE1 homolog, signal transducing adapter molecule 1, signal transducing adaptor molecule (SH3 domain and ITAM motif) 1, STAM1STAM-1
Gene Symbols STAM
Host Species Mouse
Immunogen This antibody was developed against a synthetic peptide corresponding to amino acids 149-165 of human STAM.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8027
Test Specificity A 59 kDa band is observed.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.