Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STAMP2/STEAP4 Antibody (418714), PerCP, Novus Biologicals™
SDP

Catalog No. NB369557 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB369557 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB369557 Supplier Novus Biologicals Supplier No. FAB4626C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

STAMP2/STEAP4 Monoclonal specifically detects STAMP2/STEAP4 in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen STAMP2/STEAP4
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 418714
Conjugate PerCP
Dilution Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias EC 1.16.1, EC 1.16.1.-, FLJ23153, Six-transmembrane epithelial antigen of prostate 4, SixTransMembrane protein of prostate 2, STAMP2DKFZp666D049, STEAP family member 4, TIARPsix transmembrane prostate protein 2, TNFAIP9, Tumor necrosis factor, alpha-induced protein 9metalloreductase STEAP4, tumor necrosis-alpha-induced adipose-related protein
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human STAMP2, Met1-His459, Accession # EAW76907
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 79689
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.