Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stanbio™ Bilirubin Liquid Reagent for Diagnostic Set
SDP

Catalog No. sb0240080 Shop All Stanbio Laboratory Products
Click to view available options
Quantity:
250 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Reagent liquid for Stanbio™ Diagnostic Set

  • Assay: Bilirubin
  • Assay method: Modified Walters-Gerarde
  • 20mg/L
  • 250mL
  • Working stability: 7 days

Specifications

Content And Storage 2°C to 8°C
Type Bilirubin Total LiquiColor™ Test
For Use With (Application) Modified walters-gerarde methodology
Quantity 250 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.