Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Stanbio™ Glucose Liquid Reagent for Diagnostic Set

Click to view available options
Quantity:
250 mL
Description
- Assay: Glucose
- Assay method: Colorimetric (GOD/POD)
- Linear range: 400mg/dL
- Size: 250mL
- Working stability: Until expiration date
Specifications
Specifications
Content And Storage | 2°C to 8°C |
Type | Glucose LiquiColor™ Test (Enzymatic) |
For Use With (Application) | Trinder methodology, colorimetric |
Quantity | 250 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction