Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Stanbio™ Sirrus™ General Chemistry Reagents

Click to view available options
Quantity:
100 Tests
125 Tests
90 Tests
Type:
ALT/SGPT Liqui-UV™ Test
AST/SGOT Liqui-UV™ Test
Albumin LiquiColor™ Test
Alkaline Phosphatase LiquiColor™ Test
Bicarbonate LiquiColor™ (CO2) Test
CK Liqui-UV™ Test
Calcium (Arsenazo) LiquiColor™ Test
Cholesterol LiquiColor™ Test
Creatinine LiquiColor™ Test
Direct Bilirubin LiquiColor™ Test
Direct HDL-Cholesterol LiquiColor™ Test
Glucose Liqui-UV™ Test
LDH Liqui-UV™ Test
Phosphorus Liqui-UV™ Test
Total Bilirubin LiquiColor™ Test
Total Protein LiquiColor™ Test
Urea Nitrogen Liqui-UV™ Test
Uric Acid LiquiColor™ Test
Sample Size:
12 μL
20 μL
3 μL
4 μL
4.5 μL
5 μL
6 μL
Description
- For use with Stanbio Sirrus Clinical Analyzers, Cat. Nos. SBG-550-0001, SBG-500-0001
Specifications
Specifications
Sample Type | CSF, Plasma, Serum, Urine |
Type | Glucose Liqui-UV™ Test |
For Use With (Application) | Clinical chemistry, proteins and therapeutic drug monitoring |
Quantity | 100 Tests |
Sample Size | 3 μL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction