Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stanbio™ Sirrus™ General Chemistry Reagents
SDP

Catalog No. SBS2930600 Shop All Stanbio Laboratory Products
Click to view available options
Quantity:
100 Tests
125 Tests
90 Tests
Type:
ALT/SGPT Liqui-UV™ Test
AST/SGOT Liqui-UV™ Test
Albumin LiquiColor™ Test
Alkaline Phosphatase LiquiColor™ Test
Bicarbonate LiquiColor™ (CO2) Test
CK Liqui-UV™ Test
Calcium (Arsenazo) LiquiColor™ Test
Cholesterol LiquiColor™ Test
Creatinine LiquiColor™ Test
Direct Bilirubin LiquiColor™ Test
Direct HDL-Cholesterol LiquiColor™ Test
Glucose Liqui-UV™ Test
LDH Liqui-UV™ Test
Phosphorus Liqui-UV™ Test
Total Bilirubin LiquiColor™ Test
Total Protein LiquiColor™ Test
Urea Nitrogen Liqui-UV™ Test
Uric Acid LiquiColor™ Test
Sample Size:
3 μL
4 μL
4.5 μL
5 μL
6 μL
12 μL
20 μL
This item is not returnable. View return policy

Catalog No. SBS2930600

Supplier: Stanbio™ S2930600

This item is not returnable. View return policy

  • For use with Stanbio Sirrus Clinical Analyzers, Cat. Nos. SBG-550-0001, SBG-500-0001

Specifications

Sample Type Plasma, Serum
Type ALT/SGPT Liqui-UV™ Test
For Use With (Application) Clinical chemistry, proteins and therapeutic drug monitoring
Quantity 100 Tests
Sample Size 12 μL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.