Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stanbio™ Uric Acid Liquid Reagent for Diagnostic Set
SDP

Catalog No. sb1045430 Shop All Stanbio Laboratory Products
Click to view available options
Quantity:
30 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Reagent liquid for Stanbio™ Diagnostic Set

  • Assay: Uric Acid
  • Assay method: Trinder Methodology (Enzymatic)
  • Linear range: 20mg/dL
  • Size: 30mL
  • Working stability: Until 60 days old

Specifications

Content And Storage 2°C to 8°C
Type Uric Acid LiquiColor™ Test (Enzymatic)
For Use With (Application) Trinder methodology
Quantity 30 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.