Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bal Supply Standard Taper Joint Clips
Bal Supply standard taper joint clips with superior chemical and temperature resistance
$55.70 - $83.10
Specifications
Quantity | 10/Pk. |
---|---|
Type | Standard Taper Clamp |
Material | Stainless Steel |
For Use With (Equipment) | Standard Taper Joints |
Color | Silver |
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
13-684-160
|
Bal Supply
203414 |
14 |
Case of 10 for $55.70
|
|
|||||
13-684-161
|
Bal Supply
203419 |
19 |
Case of 10 for $68.40
|
|
|||||
13-684-162
|
Bal Supply
203424 |
24 |
Case of 10 for $74.30
|
![]() |
|||||
13-684-163
|
Bal Supply
203429 |
29 |
Case of 10 for $83.10
|
|
|||||
Description
- Nickel Plated Steel
- Can withstand temperatures up to 932°F
- For Standard Taper Joints
Specifications
10/Pk. | |
Stainless Steel | |
Silver |
Standard Taper Clamp | |
Standard Taper Joints |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title