Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Stanniocalcin 2 Mouse anti-Human, Mouse, Clone: 4H1E7, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | Stanniocalcin 2 |
---|---|
Clone | 4H1E7 |
Concentration | 1 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6217
![]() ![]() |
Proteintech Group Inc
600631IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Has an anti-hypocalcemic action on calcium and phosphate homeostasis.Specifications
Stanniocalcin 2 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
stanniocalcin 2, Stanniocalcin related protein, STC 2, STC related protein, STC2, STCRP | |
STC2 | |
IgG1 | |
Caprylic Acid Precipitation | |
Antibody |
4H1E7 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse | |
O76061, O88452 | |
20856, 8614 | |
Stanniocalcin 2 Fusion Protein Ag0359 | |
Primary | |
-20°C | |
STC2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title