Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Staphylococcus Aureus Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10064499
Description
Staphylococcus Aureus Polyclonal specifically detects Staphylococcus Aureus in Bacteria samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.Specifications
Staphylococcus Aureus | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.2) with 0.1% Sodium Azide | |
Rabbit | |
IgG purified | |
RUO | |
Reacts with soluble and structural antigens of the whole bacterium. It is not absorbed for related microorganisms. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
4.0 mg/mL | |
Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunohistochemistry-Frozen | |
S. aureus | |
Staphylococcus aureus, ATCC 27660 | |
0.5 mL | |
Primary | |
Bacteria | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction