Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Staphylococcus Aureus Antibody - BSA Free, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.50 - $426.00

Specifications

Antigen Staphylococcus Aureus
Concentration 4.0 mg/mL
Dilution Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunohistochemistry-Frozen
Applications Immunohistochemistry (Frozen), Immunocytochemistry, Immunofluorescence
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB017589
SDP
View Documents
Novus Biologicals
NB100644990.1ML
0.1 mL
Each for $206.50
Only null left
Add to Cart
 
NB10064499
SDP
View Documents
Novus Biologicals
NB10064499
0.5 mL
Each for $426.00
Only null left
Add to Cart
 
Description

Description

Staphylococcus Aureus Polyclonal specifically detects Staphylococcus Aureus in Bacteria samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.
Specifications

Specifications

Staphylococcus Aureus
Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunohistochemistry-Frozen
Polyclonal
Purified
RUO
PBS (pH 7.2) with 0.1% Sodium Azide
Staphylococcus aureus, ATCC 27660
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
4.0 mg/mL
Immunohistochemistry (Frozen), Immunocytochemistry, Immunofluorescence
Unconjugated
Rabbit
Bacteria
S. aureus
IgG
IgG purified
Reacts with soluble and structural antigens of the whole bacterium. It is not absorbed for related microorganisms.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.