Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Staphylococcus Aureus Enterotoxin B Antibody (S222), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB359706 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB359706 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB359706 Supplier Novus Biologicals Supplier No. NB1108272JF549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Staphylococcus Aureus Enterotoxin B Monoclonal antibody specifically detects Staphylococcus Aureus Enterotoxin B in Bacteria samples. It is validated for Immunoassay, ELISA

Specifications

Antigen Staphylococcus Aureus Enterotoxin B
Applications Immunoassay, ELISA
Classification Monoclonal
Clone S222
Conjugate Janelia Fluor 549
Dilution Immunoassay, Sandwich ELISA
Formulation 50mM Sodium Borate
Gene Alias S. aureus
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of X63-Ag8-653 myeloma cells with spleen cells of Balb/c mice immunized with recombinant S. aureus enterotoxin B expressed in E. coli.
Purification Method Protein G purified
Quantity 0.2 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.