Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Staphylococcus Aureus Enterotoxin B Antibody (S643), HRP, Novus Biologicals™
SDP

Catalog No. NB10073025H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10073025H 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10073025H Supplier Novus Biologicals Supplier No. NB10073025H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Staphylococcus Aureus Enterotoxin B Monoclonal specifically detects Staphylococcus Aureus Enterotoxin B in Bacteria samples. It is validated for Sandwich ELISA.

Specifications

Antigen Staphylococcus Aureus Enterotoxin B
Applications Sandwich ELISA
Classification Monoclonal
Clone S643
Conjugate HRP
Dilution ELISA, Sandwich ELISA
Formulation PBS with No Preservative
Gene Alias S. aureus
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of X63-Ag8-653 myeloma cells with spleen cells of Balb/c mice immunized with recombinant S. aureus enterotoxin B expressed in E. coli.
Purification Method Protein G purified
Quantity 0.2 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Staphylococcus Aureus Enterotoxin B. This does not cross react with S. aureus enterotoxins A, C, D and E.
Target Species Bacteria
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.