Learn More
Staphylococcus aureus Monoclonal Antibody (Staph11-232.3), Invitrogen™
Mouse Monoclonal Antibody
$450.00 - $1108.00
Specifications
Antigen | Staphylococcus aureus |
---|---|
Clone | Staph11-232.3 |
Concentration | 1.4 mg/mL |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA545988
|
Invitrogen™
MA545988 |
100 μg |
Each for $450.00
|
|
|||||
01-671-939
|
Invitrogen™
MA110708 |
500 μg |
Each for $641.50
|
|
|||||
PIMA546113
|
Invitrogen™
MA546113 |
1 mg |
Each for $1,108.00
|
|
|||||
Description
MA1-10708 detects Staphylococcus aureus from Staph. Aureus and Staph. epidermidis samples. This antibody does not cross-react with Streptococcus sp., Mycoplasma sp. or Corynebacteria sp. MA1-10708 has been successfully used in ELISA applications. The MA1-10708 immunogen is UV-inactivated Staphylococcus aureus cells (ATCC #29740).
Staphylococcus aureus is a type of bacteria that commonly lives on the skin and in the nasal passages of healthy individuals. However, it can also cause a range of infections, from minor skin infections to more serious conditions such as pneumonia, sepsis, and endocarditis.Specifications
Staphylococcus aureus | |
1.4 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with no preservative; pH 7.4 | |
UV-inactivated Staphylococcus aureus cells. | |
Primary | |
4°C, do not freeze |
Staph11-232.3 | |
ELISA | |
Unconjugated | |
Mouse | |
Bacteria | |
MRSA; s aureus; S. aureus; staphylococcus aureus methicillin-resistant | |
IgG3 | |
PEG precipitation | |
Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.