Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StAR Antibody (STAR/2154), Biotin, Novus Biologicals™
SDP

Catalog No. NB008757 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB008757 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB008757 Supplier Novus Biologicals Supplier No. NBP275742B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

StAR Monoclonal specifically detects StAR in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen StAR
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone STAR/2154
Conjugate Biotin
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS with 0.05% Sodium Azide
Gene Alias cholesterol trafficker, mitochondrial steroid acute regulatory protein, StAR, StARD1, STARD1StAR-related lipid transfer (START) domain containing 1, START domain containing 1, START domain-containing protein 1, steroid acute regulatory protein, steroidogenic acute regulator, steroidogenic acute regulatory protein, steroidogenic acute regulatory protein, mitochondrial
Gene Symbols STAR
Host Species Mouse
Immunogen Recombinant fragment (around aa 39-108) of human StAR protein (exact sequence is proprietary) (Uniprot: P49675)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Test Specificity Steroidogenic Acute Regulatory Protein (STAR) controls the rate-limiting step of steroidegenesis by translocating cholesterol from the outer mitochondrial membrane to the inner membrane where it is later cleaved to pregnenolone. It is primarily present in steroid-producing cells, including Leydig cells in the testis, theca cells and luteal cells in the ovary and adrenal cells in the adrenal cortex. Due to low levels of pregnenolone, seminomas and Leydig cell tumors display no specific STAR staining. Therefore, STAR antibody may assist in differentiating sex cord stromal tumors (SCST), seminomas and embryonal carcinomas.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.