Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STARD10 Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB336767 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB336767 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB336767 Supplier Novus Biologicals Supplier No. NBP278818PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

STARD10 Polyclonal antibody specifically detects STARD10 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen STARD10
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PE
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias Antigen NY-CO-28, CGI-52, MGC14401, NY-CO-28, PCTP2, PCTP-L, PCTP-like protein, Serologically defined colon cancer antigen 28SDCCAG28, StARD10, StAR-related lipid transfer (START) domain containing 10, StAR-related lipid transfer protein 10, START domain containing 10, START domain-containing protein 10
Host Species Rabbit
Immunogen Full length recombinant human STARD10 protein (amino acids 1-291) [UniProt Q9Y365]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10809
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.