Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stargazin Antibody (S245-36), DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP222411R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222411R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222411R Supplier Novus Biologicals Supplier No. NBP222411R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Stargazin Monoclonal specifically detects Stargazin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Stargazin
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone S245-36
Conjugate DyLight 550
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias calcium channel, voltage-dependent, gamma subunit 2, FLJ41437, MGC138502, MGC138504, Neuronal voltage-gated calcium channel gamma-2 subunit, stargazin, voltage-dependent calcium channel gamma-2 subunit
Gene Symbols CACNG2
Host Species Mouse
Immunogen Fusion protein amino acids 203-323 (Cytoplasmic C-terminus) of rat TARPGamma2
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 10369
Test Specificity Detects approx 35-55kDa. Does not cross-react with TARPGamma3.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.