Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STAT1 Antibody (246523) [Biotin], Novus Biologicals™
SDP

Catalog No. NB399867 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB399867 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB399867 Supplier Novus Biologicals Supplier No. FAB1490B
Add to Cart
Add to Cart

Rat Monoclonal Antibody

STAT1 Monoclonal specifically detects STAT1 in Human samples. It is validated for CyTOF, Flow Cytometry (Intracellular Staining), Immunocytochemistry.

Specifications

Antigen STAT1
Applications CyTOF, Flow Cytometry (Intracellular Staining), Immunocytochemistry
Classification Monoclonal
Clone 246523
Conjugate Biotin
Dilution Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation PBS
Gene Alias DKFZp686B04100, ISGF-3,91kD, signal transducer and activator of transcription 1, 91kDa, signal transducer and activator of transcription-1
Host Species Rat
Immunogen E. coli-derived recombinant human STAT1, Met1-Gln194, Accession # P42224
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Breast Cancer, Cancer, Myeloid derived Suppressor Cell, Signal Transduction, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 6772
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.