Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STAT1 Antibody, Novus Biologicals™
SDP

Catalog No. NB10056314 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056314 0.1 mg
NB10056314T 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056314 Supplier Novus Biologicals Supplier No. NB10056314
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

STAT1 Polyclonal specifically detects STAT1 in Canine, Human, Mouse, Pig, Primate samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen STAT1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 μg/mL, Immunohistochemistry-Paraffin
Formulation PBS containing 0.2% gelatin with 0.05% Sodium Azide
Gene Accession No. NP_009330.1
Gene Alias DKFZp686B04100, ISGF-3,91kD, signal transducer and activator of transcription 1, 91kDa, signal transducer and activator of transcription-1
Gene Symbols STAT1
Host Species Rabbit
Immunogen A synthetic peptide sequence corresponding to amino acids between 700 and 750 of human Stat1 was used as immunogen. This sequence is 100% conserved between Stat1 alpha and Stat1 beta.
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Signal Transduction, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 6772
Test Specificity This antibody is able to detect both the alpha form (p91, ∼91 kD) and the beta form (p84, ∼84 kD) of STAT1.
Target Species Canine, Mouse, Human, Mouse, Pig, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.