Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STAT1 Antibody (OTI15H3), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB274377A48 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB274377A48 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB274377A48 Supplier Novus Biologicals Supplier No. NBP274377AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

STAT1 Monoclonal specifically detects STAT1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen STAT1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI15H3
Conjugate Alexa Fluor 488
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias DKFZp686B04100, ISGF-3,91kD, signal transducer and activator of transcription 1, 91kDa, signal transducer and activator of transcription-1
Gene Symbols STAT1
Host Species Mouse
Immunogen Full-length protein expressed in 293T cell transfected with human STAT1 expression vector.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Apoptosis, Breast Cancer, Cancer, Myeloid derived Suppressor Cell, Phospho Specific, Signal Transduction, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 6772.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.