Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STAT1 Antibody (SM1), Biotin, Novus Biologicals™
SDP

Catalog No. NB500514B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500514B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB500514B Supplier Novus Biologicals Supplier No. NB500514B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

STAT1 Monoclonal specifically detects STAT1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen STAT1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SM1
Conjugate Biotin
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS with 0.05% Sodium Azide
Gene Alias DKFZp686B04100, ISGF-3,91kD, signal transducer and activator of transcription 1, 91kDa, signal transducer and activator of transcription-1
Gene Symbols STAT1
Host Species Mouse
Immunogen STAT1 peptide sequence 721-733 (DNLLPMSPEEFDE).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Breast Cancer, Cancer, Myeloid derived Suppressor Cell, Phospho Specific, Signal Transduction, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 6772
Test Specificity The antibody SM1 recognizes an epitope included within amino acids 721-733 of STAT1, a 91 kDa transcriptional factor involved in a variety of systems including antiviral responses and interferon alpha (IFN-alpha) and gamma (IFN-gamma) signal transduction.
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.