Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STAT1, p Tyr701 Antibody (1086B), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB363835 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB363835 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB363835 Supplier Novus Biologicals Supplier No. FAB2894E
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

STAT1 Monoclonal specifically detects STAT1 in Human samples. It is validated for Flow Cytometry (Intracellular Staining), Western Blot.

Specifications

Antigen STAT1
Applications Flow Cytometry (Intracellular Staining), Western Blot
Classification Monoclonal
Clone 1086B
Conjugate DyLight 405
Dilution Western Blot, Intracellular Staining by Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias DKFZp686B04100, ISGF-3,91kD, signal transducer and activator of transcription 1, 91kDa, signal transducer and activator of transcription-1
Host Species Rabbit
Immunogen Phosphopeptide containing the human STAT1 Y701 site, Accession # NP009330
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Breast Cancer, Cancer, Myeloid derived Suppressor Cell, Phospho Specific, Signal Transduction, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 6772
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.