Learn More
Description
Specifications
Specifications
| Antigen | STAT1 |
| Applications | Flow Cytometry (Intracellular Staining), Western Blot |
| Classification | Monoclonal |
| Clone | 1086B |
| Conjugate | DyLight 594 |
| Dilution | Western Blot, Intracellular Staining by Flow Cytometry |
| Formulation | 50mM Sodium Borate |
| Gene Alias | DKFZp686B04100, ISGF-3,91kD, signal transducer and activator of transcription 1, 91kDa, signal transducer and activator of transcription-1 |
| Host Species | Rabbit |
| Immunogen | Phosphopeptide containing the human STAT1 Y701 site, Accession # NP009330 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
