Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STAT1, p Tyr701 Antibody, Novus Biologicals™
SDP

Catalog No. NBP177825 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP177825 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP177825 Supplier Novus Biologicals Supplier No. NBP177825
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

STAT1 Polyclonal specifically detects STAT1 in Human, Mouse samples. It is validated for Western Blot, ELISA.

Specifications

Antigen STAT1 (p Tyr701)
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 2.3 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, ELISA 1:25000
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Gene Accession No. NP_033309, Q99K94
Gene Alias DKFZp686B04100, ISGF-3,91kD, signal transducer and activator of transcription 1, 91kDa, signal transducer and activator of transcription-1
Gene Symbols STAT1
Host Species Rabbit
Immunogen This Protein A purified STAT1 [p Tyr701] Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to residues surrounding Y701 in the mouse STAT1 protein. (Uniprot: Q99K94)
Molecular Weight of Antigen 83.11 kDa
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis, Breast Cancer, Cancer, Phospho Specific, Signal Transduction, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 6772
Test Specificity This antibody is specific for mouse STAT1 protein phosphorylated at Y701. A BLAST analysis was used to suggest cross-reactivity with STAT1 from mouse and rat based on 100% homology with the immunizing sequence. Partial reactivity is expected against STAT1 from human sources as 90% homology is noted. Cross-reactivity with STAT1 from other sources has not been determined.
Target Species Human, Mouse
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.