Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STAT2 Antibody (STAT2/2650), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB385752 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB385752 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB385752 Supplier Novus Biologicals Supplier No. NBP308587AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

STAT2 Monoclonal antibody specifically detects STAT2 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen STAT2
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone STAT2/2650
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias interferon alpha induced transcriptional activator, ISGF-3, MGC59816, p113, signal transducer and activator of transcription 2, signal transducer and activator of transcription 2, 113kD, signal transducer and activator of transcription 2, 113kDa, STAT113
Host Species Mouse
Immunogen Recombinant full-length human STAT2 protein (Uniprot: P52630)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Signal Transduction, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 6773
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.